Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • DLC1 Antibodies

          Abnova

          DLC1 Polyclonal Antibody, MaxPab™

          View all (15) DLC1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DLC1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          DLC1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DLC1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DLC1 Antibody (H00010395-B01P) in WB

          Western Blot analysis of DLC1 expression in transfected 293T cell line by DLC1 MaxPab polyclonal antibody. Lane 1: DLC1 transfected lysate(168.08 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DLC1 Antibody in Western Blot (WB)

          Product Details

          H00010395-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DLC1 (NP_872584.1, 1 a.a. approximately 1528 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSVAIRKRSW EEHVTHWMGQ PFNSDDRNTA CHHGLVADSL QASMEKDATL NVDRKEKCVS LPDCCHGSEL RDFPGRPMGH LSKDVDENDS HEGEDQFLSL EASTETLVHV SDEDNNADLC LTDDKQVLNT QGQKTSGQHM IQGAGSLEKA LPIIQSNQVS SNSWGIAGET ELALVKESGE RKVTDSISKS LELCNEISLS EIKDAPKVNA VDTLNVKDIA PEKQLLNSAV IAQQRRKPDP PKDENERSTC NVVQDEFLDT PCTNRGLPLL KTDFGSCLLQ PPSCPNGMSA ENGLEKSGFS QHQNKSPPKV KAEDGMQCLQ LKETLATQEP TDNQVRLRKR KEIREDRDRA RLDSMVLLIM KLDQLDQDIE NALSTSSSPS GTPTNLRRHV PDLESGSESG ADTISVNQTR VNLSSDTEST DLPSSTPVAN SGTKPKTTAI QGISEKEKAE IEAKEACDWL RATGFPQYAQ LYEDFLFPID ISLVKREHDF LDRDAIEALC RRLNTLNKCA VMKLEISPHR KRSDDSDEDE PCAISGKWTF QRDSKRWSRL EEFDVFSPKQ DLVPGSPDDS HPKDGPSPGG TLMDLSERQE VSSVRSLSST GSLPSHAPPS EDAATPRTNS VISVCSSSNL AGNDDSFGSL PSPKELSSFS FSMKGHEKTA KSKTRSLLKR MESLKLKSSH HSKHKAPSKL GLIISGPILQ EGMDEEKLKQ LNCVEISALN GNRINVPMVR KRSVSNSTQT SSSSSQSETS SAVSTPSPVT RTRSLSACNK RVGMYLEGFD PFNQSTFNNV VEQNFKNRES YPEDTVFYIP EDHKPGTFPK ALTNGSFSPS GNNGSVNWRT GSFHGPGHIS LRRENSSDSP KELKRRNSSS SMSSRLSIYD NVPGSILYSS SGDLADLENE DIFPELDDIL YHVKGMQRIV NQWSEKFSDE GDSDSALDSV SPCPSSPKQI HLDVDNDRTT PSDLDSTGNS LNEPEEPSEI PERRDSGVGA SLTRSNRHRL RWHSFQSSHR PSLNSVSLQI NCQSVAQMNL LQKYSLLKLT ALLEKYTPSN KHGFSWAVPK FMKRIKVPDY KDRSVFGVPL TVNVQRTGQP LPQSIQQAMR YLRNHCLDQV GLFRKSGVKS RIQALRQMNE GAIDCVNYEG QSAYDVADML KQYFRDLPEP LMTNKLSETF LQIYQYVPKD QRLQAIKAAI MLLPDENREV LQTLLYFLSD VTAAVKENQM TPTNLAVCLA PSLFHLNTLK RENSSPRVMQ RKQSLGKPDQ KDLNENLAAT QGLAHMIAEC KKLFQVPEEM SRCRNSYTEQ ELKPLTLEAL GHLGNDDSAD YQHFLQDCVD GLFKEVKEKF KGWVSYSTSE QAELSYKKVS EGPPLRLWRS VIEVPAVPEE ILKRLLKEQH LWDVDLLDSK VIEILDSQTE IYQYVQNSMA PHPARDYVVL RTWRTNLPKG ACALLLTSVD HDRAPVVGVR VNVLLSRYLI EPCGPGKSKL TYMCRVDLRG HMPEWYTKSF GHLCAAEVVK IRDSFSNQNT ETKDTKSR

          Target Information

          This gene encodes a GTPase-activating protein that is a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins. GAP family proteins participate in signaling pathways that regulate cell processes involved in cytoskeletal changes. This gene functions as a tumor suppressor gene in a number of common cancers, including prostate, lung, colorectal, and breast cancers. Multiple transcript variants due to alternative promoters and alternative splicing have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ARHGAP 7; Deleted in liver cancer 1 protein; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC 1; DLC-1; FLJ21120; HP protein; p122 RhoGAP; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; STARD 12; StARD12; START domain-containing protein 12; transfer protein 12

          View more View less

          Gene Aliases: ARHGAP7; DLC1; HP; KIAA1723; p122-RhoGAP; STARD12

          View more View less

          UniProt ID: (Human) Q96QB1

          View more View less

          Entrez Gene ID: (Human) 10395

          View more View less

          Function(s)
          GTPase activator activity protein binding lipid binding SH2 domain binding
          Process(es)
          neural tube closure heart morphogenesis apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process signal transduction negative regulation of cell proliferation regulation of cell shape hindbrain morphogenesis actin cytoskeleton organization negative regulation of cell migration forebrain development regulation of actin cytoskeleton organization negative regulation of Rho protein signal transduction positive regulation of protein dephosphorylation positive regulation of GTPase activity focal adhesion assembly regulation of small GTPase mediated signal transduction negative regulation of stress fiber assembly negative regulation of focal adhesion assembly positive regulation of execution phase of apoptosis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0