Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MVLLIMKLDQ LDQDIENALS TSSSPSGTPT NLRRHVPDLE SGSESGADTI SVNQTRVNLS SDTESTDLPS STPVANSGTK PKTTAIQGI
Antigen sequence identity: Mouse 79%; Rat 28%
This gene encodes a GTPase-activating protein that is a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins. GAP family proteins participate in signaling pathways that regulate cell processes involved in cytoskeletal changes. This gene functions as a tumor suppressor gene in a number of common cancers, including prostate, lung, colorectal, and breast cancers. Multiple transcript variants due to alternative promoters and alternative splicing have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARHGAP 7; Deleted in liver cancer 1 protein; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC 1; DLC-1; FLJ21120; HP protein; p122 RhoGAP; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; STARD 12; StARD12; START domain-containing protein 12; transfer protein 12
Gene Aliases: ARHGAP7; DLC1; HP; KIAA1723; p122-RhoGAP; STARD12
UniProt ID: (Human) Q96QB1
Entrez Gene ID: (Human) 10395
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support