Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRPCANGAT CLDGINRFSC LCPEGFAGRF CTINLDDCAS RPCQRGARCR DRVHDFDCLC PSGYGGKTCE LVLPVPDPPT TVDTPLGPTS AVVVPATGPA PHSAGAGLLR ISVKEVVRRQ EAGLGEPSLV ALVVFGALTA ALVLATVLLT LRAWRRGVCP PGPCCYPAPH YAPACQDQEC QVSMLPAGLP LPRDLPPEPG KTTAL (180-383 aa encoded by BC006425)
Regulates adipogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: delta-like 2 homolog; DLK-2; EGF-like protein 9; EGF-like-domain, multiple 9; Endothelial cell-specific protein S-1; Epidermal growth factor-like protein 9; Protein delta homolog 2
Gene Aliases: AI413481; DLK-2; DLK2; EGFL9; UNQ2903/PRO28633
UniProt ID: (Human) Q6UY11, (Mouse) Q8K1E3, (Rat) D3ZUK3
Entrez Gene ID: (Human) 65989, (Mouse) 106565, (Rat) 316232
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support