Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPSLAGPPFG AEAAGSGYPG PLDLRRPMRT VPGPLFTDFV RPLNINPDRA LGPEYPGGSS MHPYCPFPLG YLDAPPGVPL QQGFRHVSRS QYQGGGLVSE PGGDFQPSYY LPPPPPPLPP LPPLPPQPQF LPPGYLSALH FLPPPPPPPP PSSFSLTVLF DTDKENTDDQ DAEVLSGEPS QPSSQEQSD
DMRTB1 gene ontology annotations related to this gene include DNA-binding transcription factor activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DMRT-like family B with proline-rich C-terminal 1; DMRT-like family B with proline-rich C-terminal, 1; Doublesex- and mab-3-related transcription factor B1
Gene Aliases: DMRTB1
UniProt ID: (Human) Q96MA1
Entrez Gene ID: (Human) 63948
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support