Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • DNAI1 Antibodies

          Abnova

          DNAI1 Polyclonal Antibody, MaxPab™

          View all (10) DNAI1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DNAI1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          DNAI1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DNAI1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DNAI1 Antibody (H00027019-B01P) in WB

          Western Blot analysis of DNAI1 expression in transfected 293T cell line by DNAI1 MaxPab polyclonal antibody. Lane 1: DNAI1 transfected lysate(76.89 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DNAI1 Antibody in Western Blot (WB)

          Product Details

          H00027019-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DNAI1 (NP_036276.1, 1 a.a. approximately 699 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MIPASAKAPH KQPHKQSISI GRGTRKRDED SGTEVGEGTD EWAQSKATVR PPDQLELTDA ELKEEFTRIL TANNPHAPQN IVRYSFKEGT YKPIGFVNQL AVHYTQVGNL IPKDSDEGRR QHYRDELVAG SQESVKVISE TGNLEEDEEP KELETEPGSQ TDVPAAGAAE KVTEEELMTP KQPKERKLTN QFNFSERASQ TYNNPVRDRE CQTEPPPRTN FSATANQWEI YDAYVEELEK QEKTKEKEKA KTPVAKKSGK MAMRKLTSME SQTDDLIKLS QAAKIMERMV NQNTYDDIAQ DFKYYDDAAD EYRDQVGTLL PLWKFQNDKA KRLSVTALCW NPKYRDLFAV GYGSYDFMKQ SRGMLLLYSL KNPSFPEYMF SSNSGVMCLD IHVDHPYLVA VGHYDGNVAI YNLKKPHSQP SFCSSAKSGK HSDPVWQVKW QKDDMDQNLN FFSVSSDGRI VSWTLVKRKL VHIDVIKLKV EGSTTEVPEG LQLHPVGCGT AFDFHKEIDY MFLVGTEEGK IYKCSKSYSS QFLDTYDAHN MSVDTVSWNP YHTKVFMSCS SDWTVKIWDH TIKTPMFIYD LNSAVGDVAW APYSSTVFAA VTTDGKAHIF DLAINKYEAI CNQPVAAKKN RLTHVQFNLI HPIIIVGDDR GHIISLKLSP NLRKMPKEKK GQEVQKGPAV EIAKLDKLLN LVREVKIKT

          Target Information

          Eukaryotic cells rely on actin and microtubule-based protein "motors" to generate intracellular movements. These protein "motors" contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (actin in the case of myosin family and microtubules in the case of the kinesin family and dyneins). Dynein has been implicated in cytoplasmic motile functions, including chromosomal movement, retrograde organelle and axonal transport, the endocytic pathway, and the organization of the Golgi apparatus. In all cell types, dynein has the same basic structures and is composed of two or three distinct heavy chains (~450 kDa), three intermediate chains (70-125 kDa), and at least four light chains (15-25 kDa).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Axonemal dynein intermediate chain 1; Dynein axonemal intermediate chain 1; dynein, axonemal, intermediate chain 1; dynein, axonemal, intermediate polypeptide 1; ICS; immotile cilia syndrome 1; MGC26204; RP11-296L22.2; testis tissue sperm-binding protein Li 87P

          View more View less

          Gene Aliases: CILD1; DIC1; DNAI1; ICS1; PCD

          View more View less

          UniProt ID: (Human) Q9UI46

          View more View less

          Entrez Gene ID: (Human) 27019

          View more View less

          Function(s)
          motor activity protein binding
          Process(es)
          cilium movement determination of left/right symmetry sperm motility outer dynein arm assembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0