Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HDJ2 Antibodies

          Proteintech

          DNAJA1 Monoclonal Antibody (1B10A10), CoraLite® Plus 488

          View all (15) HDJ2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DNAJA1 Monoclonal Antibody (1B10A10), CoraLite® Plus 488

          • Antibody Testing Data (2)
          DNAJA1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DNAJA1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DNAJA1 Antibody (CL488-67184) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite® Plus 488 DNAJA1 antibody (CL488-67184, Clone: 1B10A10 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DNAJA1 Antibody in Immunocytochemistry (ICC/IF)
          DNAJA1 Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          CL488-67184

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgM

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1B10A10

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • CoraLite 594
          • Request custom conjugation

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Purification

          Euglobulin precipitation

          Storage buffer

          PBS with 0.5% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MVKETTYYDV LGVKPNATQE ELKKAYRKLA LKYHPDKNPN EGEKFKQISQ AYEVLSDAKK RELYDKGGEQ AIKEGGAGGG FGSPMDIFDM FFGGGGRMQR ERRGKNVVHQ LSVTLEDLYN GATRKLALQK NVICDKCEGR GGKKGAVECC PNCRGTGMQI RIHQIGPGMV QQIQSVCMEC QGHGERISPK DRCKSCNGRK IVREKKILEV HIDKGMKDGQ KITFHGEGDQ EPGLEPGDII IVLDQKDHAV FTRRGEDLFM CMDIQLVEAL CGFQKPISTL DNRTIVITSH PGQIVKHGDI KCVLNEGMPI YRRPYEKGRL IIEFKVNFPE NGFLSPDKLS LLEKLLPERK EVEETDEMDQ VELVDFDPNQ ERRRHYNGEA YEDDEHHPRG GVQCQTS

          Target Information

          The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ homolog subfamily A member 1; DnaJ protein homolog 2; DnaJ-like protein 1; DnaJ-like protein 2; h HSJ-2; hDj-2; Heat shock 40 kDa protein 4; Heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ-2; Human DnaJ protein 2; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7

          View more View less

          Gene Aliases: DJ-2; DjA1; DNAJ2; DNAJA1; hDJ-2; HDJ2; HSDJ; HSJ-2; HSJ2; HSPF4; NEDD7; Rdj1

          View more View less

          UniProt ID: (Human) P31689, (Mouse) P63037, (Rat) P63036

          View more View less

          Entrez Gene ID: (Human) 3301, (Mouse) 15502, (Rat) 65028

          View more View less

          Function(s)
          G-protein coupled receptor binding protein binding ATP binding Hsp70 protein binding Tat protein binding ubiquitin protein ligase binding metal ion binding low-density lipoprotein particle receptor binding unfolded protein binding chaperone binding C3HC4-type RING finger domain binding heat shock protein binding
          Process(es)
          protein folding spermatogenesis response to heat sperm motility androgen receptor signaling pathway negative regulation of protein ubiquitination protein refolding DNA damage response, detection of DNA damage positive regulation of apoptotic process negative regulation of apoptotic process negative regulation of JUN kinase activity regulation of protein transport protein localization to mitochondrion toxin transport response to unfolded protein negative regulation of establishment of protein localization to mitochondrion negative regulation of nitrosative stress-induced intrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0