Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MGKDYYCIL GIEKGASDED IKKAYRKQAL KFHPDKNKSP QAEEKFKEVA EAYEVLSDPK KREIYDQFGE EGLKGGAGGT DGQGGTFRYT FHGDPHATFA AFFGGSNPFE IFFGRRMGGG RDSEEMEIDG DPFSAFGFSM NGYPRDRNSV GPSRLKQDPP VIHELRVSLE EIYSGCTKRM KISRKRLNAD GRSYRSEDKI LTIEIKKGWK EGTKITFPRE GDETPNSIPA DIVFIIKDKD HPKFKRDGSN IIYTAKISLR EALCGCSINV PTLDGRNIPM SVNDIVKPGM RRRIIGYGLP FPKNPDQRGD LLIEFEVSFP DTISSSSKEV LRKHLPAS (1-337 aa encoded by BC034721)
DnaJ heat shock induced proteins are from the bacterium Escherichia coli and are under the control of the htpR regulatory protein. The DnaJ proteins play a critical role in the HSP 70 chaperone machine by interacting with HSP 70 to stimulate ATP hydrolysis. The proteins contain cysteine rich regions that are composed of zinc fingers that form a peptide binding domain responsible for the chaperone function. DnaJ proteins are important mediators of proteolysis and are involved in the regulation of protein degradation, exocytosis and endocytosis. DnaJB4 (DnaJ homolog subfamily B member 4), also known as HLJ1, is expressed in skeletal muscle, heart and pancreas, and lower expression in brain, placenta and liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily B, member 4; DnaJ (Hsp40) homolog, subfamily B, member 4-like; DnaJ homolog subfamily B member 4; DnaJ-like heat shock protein 40; DNAJB 4; Heat shock 40 kDa protein 1 homolog; heat shock protein 40 homolog; HLJ 1; HSP40 homolog; Human liver DnaJ-like protein
Gene Aliases: 1700029A20Rik; 2010306G19Rik; 5730460G06Rik; DjB4; DNAJB4; DNAJW; HLJ1
UniProt ID: (Human) Q9UDY4, (Mouse) Q9D832
Entrez Gene ID: (Human) 11080, (Rat) 295549, (Mouse) 67035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support