Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: axonemal dynein light chain (hp28) homolog; Axonemal dynein light intermediate polypeptide 1; dJ423B22.5 (axonemal dynein light chain (hp28)); dynein, axonemal, light intermediate chain 1; dynein, axonemal, light intermediate polypeptide 1; hp28; Inner dynein arm light chain, axonemal; inner dynein arm, homolog of clamydomonas
Gene Aliases: 1700023A09Rik; AW049135; dJ423B22.5; DNALI1; hp28; P28
UniProt ID: (Human) O14645, (Mouse) Q8BVN8
Entrez Gene ID: (Human) 7802, (Mouse) 75563
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support