Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MHYPTALLFL ILANGAQAFR ICAFNAQRLT LAKVAREQVM DTLVRILARC DIMVLQEVVD SSGSAIPLLL RELNRFDGSG PYSTLSSPQL GRSTYMETYV YFYRSHKTQV LSSYVYNDED DVFAREPFVA QFSLPSNVLP SLVLVPLHTT PKAVEKELNA LYDVFLEVSQ HWQSKDVILL GDFNADCASL TKKRLDKLEL RTEPGFHWVI ADGEDTTVRA STHCTYDRVV LHGERCRSLL HTAAAFDFPT SFQLTEEEAL NISDHYPVEV ELKLSQAHSV QPLSLTVLLL LSLLSPQLCP AA; Positive Samples: Mouse heart; Cellular Location: Endoplasmic reticulum
DNASE1L1 encodes a deoxyribonuclease protein that shows high sequence similarity to DNase I. The encoded protein is localized to the endoplasmic reticulum and modified by N-linked glycosylation. Alternate transcriptional splice variants encoding the same protein have been observed.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deoxyribonuclease I-like 1; Deoxyribonuclease-1-like 1; DNase I-like 1; DNase X
Gene Aliases: 2310005K03Rik; AI324885; Dnase1l1; Dnase1ll; Dnasex; Dnl1ll; G4.8
UniProt ID: (Mouse) Q9D7J6, (Rat) Q2QDE7
Entrez Gene ID: (Mouse) 69537, (Rat) 363522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support