Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSRELAPLL LLLLSIHSAL AMRICSFNVR SFGESKQEDK NAMDVIVKVI KRCDIILVME IKDSNNRICP ILMEKLNRNS RRGITYNYVI SSRLGRNTYK EQYAFLYKEK LVSVKRSYHY HDYQDGDADV FSREPFVVWF QSPHTAVKDF VIIPLHTTPE TSVKEIDELV EVYTDVKHRW KAENFIFMGD FNAGCSYVPK KAWKNIRLRT DPRFVWLIGD QEDTTVKKST NCAYDRIVLR GQEIVSSVVP KSNSVFDFQK AYKLTEEEAL DVSDHFPVEF KLQSSRAFTN SKKSVTLRKK TKSKRS (1-305 aa encoded by B C015831)
This gene encodes a member of the DNase family. The protein hydrolyzes DNA, is not inhibited by actin, and mediates the breakdown of DNA during apoptosis. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deoxyribonuclease gamma; Deoxyribonuclease I-like 3; deoxyribonuclease I-like III; DNase gamma; DNase I homolog protein 2; DNase I homolog protein DHP2; DNase I-like 3; Liver and spleen DNase; LS-DNase
Gene Aliases: DHP2; DNAS1L3; DNASE1L3; LSD; SLEB16
UniProt ID: (Human) Q13609
Entrez Gene ID: (Human) 1776
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support