Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Dynamin 2 Antibodies

          Proteintech

          DNM2 Polyclonal Antibody

          2 References
          View all (16) Dynamin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DNM2 Polyclonal Antibody

          • Antibody Testing Data (7)
          DNM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DNM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DNM2 Antibody (14605-1-AP) in WB

          Human brain tissue were subjected to SDS PAGE followed by western blot with 14605-1-AP (DNM2 antibody) at dilution of 1:200 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DNM2 Antibody in Western Blot (WB)
          DNM2 Antibody in Western Blot (WB)
          DNM2 Antibody in Western Blot (WB)
          DNM2 Antibody in Western Blot (WB)
          DNM2 Antibody in Western Blot (WB)
          DNM2 Antibody in Western Blot (WB)
          DNM2 Antibody in Western Blot (WB)

          Product Details

          14605-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DNM2 Fusion Protein Ag6156 (518-866 aa encoded by BC054501)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.18 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: RRGWLTINN ISLMKGGSKE YWFVLTAESL SWYKDEEEKE KKYMLPLDNL KIRDVEKGFM SNKHVFAIFN TEQRNVYKDL RQIELACDSQ EDVDSWKASF LRAGVYPEKD QAENEDGAQE NTFSMDPQLE RQVETIRNLV DSYVAIINKS IRDLMPKTIM HLMINNTKAF IHHELLAYLY SSADQSSLME ESADQAQRRD DMLRMYHALK EALNIIGDIS TSTVSTPVPP PVDDTWLQSA SSHSPTPQRR PVSSIHPPGR PPAVRGPTPG PPLIPVPVGA AASFSAPPIP SRPGPQSVFA NSDLFPAPPQ IPSRPVRIPP GIPPGVPSRR PPAAPSRPTI IRPAEPSLLD (518-866 aa encoded by BC054501)

          Target Information

          Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Five alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Dynamin 2; Dynamin II; Dynamin UDNM; Dynamin-2

          View more View less

          Gene Aliases: b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; DNM2; DYIIAAB; DYN2; DYNII; LCCS5; Udnm

          View more View less

          UniProt ID: (Human) P50570, (Rat) P39052, (Mouse) P39054

          View more View less

          Entrez Gene ID: (Human) 1785, (Rat) 25751, (Mouse) 13430

          View more View less

          Function(s)
          GTPase activity protein binding GTP binding microtubule binding SH3 domain binding enzyme binding protein kinase binding D2 dopamine receptor binding protein complex binding phosphatidylinositol 3-kinase regulatory subunit binding WW domain binding nitric-oxide synthase binding nucleotide binding hydrolase activity
          Process(es)
          G-protein coupled receptor internalization Golgi to plasma membrane transport endocytosis receptor-mediated endocytosis spermatogenesis metabolic process response to light stimulus positive regulation of lamellipodium assembly negative regulation of transforming growth factor beta receptor signaling pathway regulation of axon extension receptor internalization transferrin transport regulation of Rac protein signal transduction response to cocaine macropinocytosis positive regulation of nitric oxide biosynthetic process positive regulation of endocytosis neuron projection morphogenesis positive regulation of phagocytosis cilium morphogenesis cellular response to carbon monoxide cellular response to X-ray cellular response to nitric oxide positive regulation of substrate adhesion-dependent cell spreading cellular response to dopamine regulation of Golgi organization positive regulation of sodium:potassium-exchanging ATPase activity negative regulation of membrane tubulation G2/M transition of mitotic cell cycle ventricular septum development regulation of transcription, DNA-templated phagocytosis signal transduction antigen processing and presentation of exogenous peptide antigen via MHC class II aorta development positive regulation of apoptotic process positive regulation of transcription, DNA-templated synaptic vesicle transport regulation of nitric-oxide synthase activity coronary vasculature development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0