Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) DOCK1, rhDOCK2, and rhDOCK5 is observed.
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Dedicator of cytokinesis3 (DOCK3), also named as MOCA and PBP, is a ~180 kDa protein involved in signaling transduction. It is a potential guanine nucleotide exchange factor (GEF) which activate some small GTPases by exchanging bound GDP for free GTP. DOCK3 is associated in Alzheimer disease tangles and regulates the accumulation of amyloid precursor protein and beta-amyloid. Overexpression of Dock3 in neural cells promotes axonal outgrowth downstream of brain-derived neurotrophic factor (BDNF) signaling. DOCK3 binds to and inactivates glycogen synthase kinase-3beta (GSK-3beta) at the plasma membrane, thereby promoting axon branching and microtubule assembly. By stimulating actin polymerization and microtubule assembly, DOCK3 plays important roles downstream of BDNF signaling in the CNS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dedicator of cytokinesis protein 3; dok-3; Modifier of cell adhesion; PBP; Presenilin-binding protein
Gene Aliases: DOCK3; KIAA0299; MOCA; PBP
UniProt ID: (Human) Q8IZD9
Entrez Gene ID: (Human) 1795
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support