Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Multiple isoforms of DOCK 7 exist due to alternative splicing events. The DOCK 7 antibody (13000-1-AP) can recognize all isoforms.
Immunogen sequence: MAGMYEAVN EVYKVLIPIH EANRDAKKLS TIHGKLQEAF SKIVHQDGKR MFGTYFRVGF YGTKFGDLDE QEFVYKEPAI TKLAEISHRL EGFYGERFGE DVVEVIKDSN PVDKCKLDPN KAYIQITYVE PYFDTYEMKD RITYFDKNYN LRRFMYCTPF TLDGRAHGEL HEQFKRKTIL TTSHAFPYIK TRVNVTHKEE IILTPIEVAI EDMQKKTQEL AFATHQDPAD PKMLQMVLQG SVGTTVNQGP LEVAQVFLSE IPSDPKLFRH HNKLRLCFKD FTKRCEDALR KNKSLIGPDQ KEYQRELERN YHRLKEALQP LINRKIPQLY KAVLPVTCHR DSFSRMSLRK MDL (447-798 aa encoded by BC016392)
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) that plays a role in axon formation and neuronal polarization. The encoded protein displays GEF activity toward RAC1 and RAC3 Rho small GTPases but not toward CDC42. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dedicator of cytokinesis protein 7; protein moonlight
Gene Aliases: 3110056M06Rik; DOCK7; EIEE23; Gm430; KIAA1771; m; ZIR2
UniProt ID: (Human) Q96N67
Entrez Gene ID: (Human) 85440, (Mouse) 67299, (Rat) 313388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support