Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MFSFEAGRR CPSGPGTFTF QTAQGNDIFQ AVETAIHRQK AQGKAGQGHD VLRADSHEGE VAEGKLPSPP GPQELLDSPP ALYAEPLDSL RIAPCPSQDS LYSDPLDSTS AQAGEGVQRK KPLYWDLYEH AQQQLLKAKL TDPKEDPIYD EPEGLAPVPP QGLYDLPREP KDAWWCQARV KEEGYELPYN PATDDYAVPP PRSTKPLLAP KPQGPAFPEP GTATGSGIKS HNSALYSQVQ KSGASGSWDC GLSRVGTDKT GVKSEGST (1-267 aa encoded by BC111055)
DOK1 (P62dok, Docking protein 1) is believed to be a mainly cytoplasmic adaptor protein which down-regulates mitogen-activated protein kinase activation, inhibits cell proliferation and transformation, and promotes cell spreading and cell migration. DOK1 is a major substrate for many tyrosine kinases including c-kit, v-abl, vFPS, EGF, and PDGF. Upon phosphorylation by kinases, DOK1 forms a complex with ras GTPase-activating protein. Phosphorylation on tyrosine residues by the insulin receptor kinase results in the negative regulation of the insulin signaling pathway. DOK1 contains a putative pleckstrin homology domain at the amino terminus and ten PXXP SH3 recognition motifs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Docking protein 1; docking protein 1, 62kDa (downstream of tyrosine kinase 1); DOK 1; Downstream of tyrosine kinase 1; MGC117395; MGC138860; p240; p62 dok; p62(dok); pp62; TLP1; TP1; TROVE domain family; TROVE1; VAULT2
Gene Aliases: DOK1; P62DOK; pp62
UniProt ID: (Human) Q99704
Entrez Gene ID: (Human) 1796
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support