Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes a member of the ring between ring fingers (RBR) protein family, and the encoded protein contains two RING-finger motifs and an in between RING fingers motif. This protein is an E3 ubiquitin ligase that is localized to Lewy bodies, and ubiquitylates synphilin-1, which is an interacting protein of alpha synuclein in neurons. The encoded protein may be involved in amyotrophic lateral sclerosis and Parkinson's disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dorfin; Double ring-finger protein; E3 ubiquitin-protein ligase RNF19A; Gametogenesis-expressed protein GEG-154; p38; protein p38 interacting with transcription factor Sp1; ring finger protein (C3HC4 type) 19; ring finger protein 19; RING finger protein 19A; ring finger protein 19A, E3 ubiquitin protein ligase; ring-IBR-ring domain containing protein Dorfin; UBCM4-interacting protein 117; ubiquitin conjugating enzyme 7 interacting protein 2; UIP117; XY body protein; XYbp
Gene Aliases: AA032313; Dorfin; Geg-154; RNF19; RNF19A; Ubce7ip2; UIP117; XYbp
UniProt ID: (Human) Q9NV58, (Mouse) P50636
Entrez Gene ID: (Human) 25897, (Mouse) 30945, (Rat) 362900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support