Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: ILGCTSPRLS KEVEAVVYLG DGRFHLESVM IANPNVPAYR YDPYSKVLSR EHYDHQRMQA ARQEAIATAR SAKSWGLILG TLGRQGSPKI LEHLESRLRA LGLSFVRLL
Diphthamide is a unique posttranslationally modified histidine found only in translation elongation factor-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-(3-amino-3-carboxypropyl)histidine synthase subunit 1; candidate tumor suppressor in ovarian cancer 1; Diphthamide biosynthesis protein 1; diphthamide biosynthesis protein 2 homolog-like 1; Diphtheria toxin resistance protein 1; diptheria toxin resistance protein required for diphthamide biosynthesis (Saccharomyces)-like 1; diptheria toxin resistance protein required for diphthamide biosynthesis-like 1; DPH-like 1; DPH1 homolog; DPH2-like 1; FLJ33211; Ovarian cancer-associated gene 1 protein; ovarian tumor suppressor candidate 1; S-adenosyl-L-methionine:L-histidine 3-amino-3-carboxypropyltransferase 1
Gene Aliases: DEDSSH; DPH1; DPH2L; DPH2L1; OVCA1
UniProt ID: (Human) Q9BZG8
Entrez Gene ID: (Human) 1801
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support