Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MADTQYILPN DIGVSSLDCR EAFRLLSPTE RLYAYHLSRA AWYGGLAVLL QTSPEAPYIY ALLSRLFRAQ DPDQLHQHAL AEGLTEEEYQ AFLVYAAGVY SNMGNYKSFG DTKFVPNLPK EKLERVILGS EAAQQHPEEV RGLWQTCGEL MFSLEPRLRH LGLGKEGITT YFSGNCTMED AKLAQDFLDS QNLSAYNTRL FKEVDGEGKP YYEVRLASVL GSEPSLDSEV TSKLKSYEFR GSPFQVTRGD YAPILQKVVE QLEKAKAYAA NSHQGQMLAQ YIESFTQGSI EAHKRGSRFW IQDKGPIVES YIGFIESYRD PFGSRGEFEG FVAVVNKAMS AKFERLVASA EQLLKELPWP PTFEKDKFLT PDFTSLDVLT FAGSGIPAGI NIPNYDDLRQ TEGFKNVSLG NVLAVAYATQ REKLTFLEED DKDLYILWKG PSFDVQVGLH ELLGHGSGKL FVQDEKGAFN FDQETVINPE TGEQIQSWYR SGETWDSKFS TIASSYEECR AESVGLYLCL HPQVLEIFGF EGADAEDVIY VNWLNMVRAG LLALEFYTPE AFNWRQAHMQ ARFVILRVLL EAGEGLVTIT PTTGSDGRPD ARVRLDRSKI RSVGKPALER FLRRLQVLKS TGDVAGGRAL YEGYATVTDA PPECFLTLRD TVLLRKESRK LIVQPNTRLE GSDVQLLEYE ASAAGLIRSF SERFPEDGPE LEEILTQLAT ADARFWKGPS EAPSGQA
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dipeptidyl aminopeptidase III; Dipeptidyl arylamidase III; Dipeptidyl peptidase 3; Dipeptidyl peptidase III; DPP III; Enkephalinase B; hCG_2016942
Gene Aliases: DPP3; DPPIII
UniProt ID: (Human) Q9NY33
Entrez Gene ID: (Human) 10072
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support