Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Positive Control - WB: human Hela whole cell, human Jurkat whole cell, human HepG2 whole cell. IHC: human rectum adenocarcinoma tissue, human ovarian serous adenocarcinoma tissue. Flow: HEL cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Belongs to the DPY19 family. There are two named isoforms. The specific function of DPY19L1 is not yet known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dpy-19-like 1; DPY-19-like protein 1; Protein C-mannosyl-transferase DPY19L1; Protein dpy-19 homolog 1
Gene Aliases: DPY19L1; GA0500; KIAA0877
UniProt ID: (Human) Q2PZI1
Entrez Gene ID: (Human) 23333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support