Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GELDDRFLD DLGLKFKTLA EVCLGQKIDI NKEIEQRQKP ATETSMNTAS HSLCEQTMVN SENTYSSGSS FPVPKSLQEA NAEKVTQEIV TERSVSSRQA QKVATPLPDP MASRNVIATE TSYVTGSTMP PTTVILGPSQ PQSLIVTERV YAPASTLVDQ PYANEGTVVV TERVIQPHGG GSNPLEGTQH LQDVPYVMVR ERESFLAPSS GVQPTLAMPN IAVGQNVTVT ERVLAPASTL QSSYQIPTEN SMTARNTTVS GAGVPGPLPD FGLEESGHSN STITTSSTRV TKHSTVQHSY S (819-1118 aa encoded by BC099655)
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARVC10; ARVD10; Cadherin family member 5; CMD1BB; desmoglein 2; Desmoglein-2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Gene Aliases: AA408168; CDHF5; D18Ertd293e; DSG2; HDGC
UniProt ID: (Human) Q14126, (Mouse) O55111
Entrez Gene ID: (Human) 1829, (Mouse) 13511, (Rat) 307562
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support