Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MIEESGNKRK TMAEKRQLFI EMRAQNFDVI RLSTYRTACK LRFVQKRCNL HLVDIWNMIE AFRDNGLNTL DHTTEISVSR LETVISSIYY QLNKRLHSTH QISVEQSISL LLNFMTAAYD SEGRGKLTVF SVKAMLATMC GGKMLDKLRY VFSQMSDSNG LMIFSKFDQF LKEVLKLPTA VFEGPSFGYT EHSVRTCFPQ QRKIMLNMFL DTMMADPPPQ CLVWLPLMHR LAHVENVFHP VECSYCRCES MMGFRYRCQQ CHNYQLCQNC FWRGHAGGPH SNQHQMKEHS SWKSPAKKLS HAISKSLGCV PTREPPHPVF PEQPEKPLDL AHIVPPRPLT NMNDTMVSHM SSGVPTPTKR LQYSQDIPSH LADEHALIAS YVARLQHCAR VLDSPSRLDE EHRLIARYAA RLAAEAGNVT RPPTDLSFNF DANKQQRQLI AELENKNREI LQEIQRLRLE HEQASQPTPE KAQQNPTLLA ELRLLRQRKD ELEQRMSALQ ESRRELMVQL EELMKLLKEE EQKQAAQATG SPHTSPTHGG GRPMPMPVRS TSAGSTPTHC PQDSLSGVGG DVQEAFAQGT RRNLRNDLLV AADSITNTMS SLVKELHSG
This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-dystrobrevin; DTN-B; dtnb {ECO:0000250|UniProtKB:O70585}; Dystrobrevin beta; dystrobrevin, beta
Gene Aliases: DTNB
UniProt ID: (Human) O60941
Entrez Gene ID: (Human) 1838, (Rat) 362715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support