Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: METNYLKRCF GNCLAQALAE VAKVRPSDPI EYLAHWLYHY RKTAKAKEEN REKKIHLQEE YDSSLKEMEM TEMLKQEEYQ IQQNCEKCHK ELTSETVSTK KTIFMQEDTN PLEKEALKQE FLPGTSSLIP GMPQQVPPSE SAGQIDQNFK MPQEINYKEA FQHEVAHEMP PGSKSPF
DYDC2 encodes a member of a family of proteins that contains a DPY30 domain. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA36D19.6; DPY30 domain-containing protein 2; MGC16186; SPS1; SRML1
Gene Aliases: DYDC2
UniProt ID: (Human) Q96IM9
Entrez Gene ID: (Human) 84332
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support