Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MSDKSELKAE LERKKQRLAQ IREEKKRKEE ERKKKETDQK KEAVAPVQEE SDLEKKRREA EALLQSMGLT PESPIVPPPM SPSSKSVSTP SEAGSQDSGD GAVGSRRGPI KLGMAKITQV DFPPREIVTY TKETQTPVMA QPKEDEEEDD DVVAPKPPIE PEEEKTLKKD EENDSKAPPH ELTEEEKQQI LHSEEFLSFF DHSTRIVERA LSEQINIFFD YSGRDLEDKE GEIQAGAKLS LNRQFFDERW SKHRVVSCLD WSSQYPELLV ASYNNNEDAP HEPDGVALVW NMKYKKTTPE YVFHCQSAVM SATFAKFHPN LVVGGTYSGQ IVLWDNRSNK RTPVQRTPLS AAAHTHPVYC VNVVGTQNAH NLISISTDGK ICSWSLDMLS HPQDSMELVH KQSKAVAVTS MSFPVGDVNN FVVGSEEGSV YTACRHGSKA GISEMFEGHQ GPITGIHCHA AVGAVDFSHL FVTSSFDWTV KLWTTKNNKP LYSFEDNADY VYDVMWSPTH PALFACVDGM GRLDLWNLNN DTEVPTASIS VEGNPALNRV RWTHSGREIA VGDSEGQIVI YDVGEQIAVP RNDEWARFGR TLAEINANRA DAEEEAATRI PA
Dyneins are multi-subunit, high molecular weight ATPases that interact with microtubules to generate force by converting the chemical energy of ATP into the mechanical energy of movement. Cytoplasmic or axonemal Dynein heavy, intermediate, light and light-intermediate chains are all components of minus end-directed motors; the complex that transports cellular cargos towards the central region of the cell. Axonemal Dynein motors contain one to three non-identical heavy chains and cause a sliding of microtubules in the axonemes of cilia and flagella in a mechanism necessary for cilia to beat and propel the cell. Cytoplasmic Dynein is an approximately 12 subunit complex of two heavy chains, two intermediate chains to anchor Dynein to its cargo, four smaller intermediate chains and several light chains. It performs functions necessary for cell survival such as organelle transport and centrosome assembly. The carboxy-terminus of Dynein is important for microtubule-dependent motility and is highly conserved, while the amino-terminal regions are more variable. Several proteins regulate Dynein activity, including Dynactin, LIS1 and nudel (nudE-like).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic dynein 1 intermediate chain 2; Cytoplasmic dynein intermediate chain 2; DH IC-2; Dynein intermediate chain 2, cytosolic; dynein, cytoplasmic 1, intermediate chain 2; dynein, cytoplasmic, intermediate polypeptide 2; testis tissue sperm-binding protein Li 66n
Gene Aliases: DIC74; DNCI2; DNCIC2; DYNC1I2; IC2
UniProt ID: (Human) Q13409
Entrez Gene ID: (Human) 1781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support