Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAPVGVEKKL LLGPNGPAVA AAGDLTSEEE EGQSLWSSIL SEVSTRARSK LPSGKNILVF GEDGSGKTTL MTKLQGAEHG KKGRGLEYLY LSVHDEDRDD HTRCNVWILD GDLYHKGLLK FAVSAESLPE TLVIFVADMS RPWTVMESLQ KWASVLREHI DKMKIPPEKM RELERKFVKD FQDYMEPEEG CQGSPQRRGP LTSGSDEENV ALPLGDNVLT HNLGIPVLVV CTKCDAVSVL EKEHDYRDEH LDFIQSHLRR FCLQYGAALI YTSVKEEKNL DLLYKYIVHK TYGFHFTTPA LVVEKDAVFI PAGWDNEKKI AILHENFTTV KPEDAYEDFI VKPPVRKLVH DKELAAEDEQ VFLMKQQSLL AKQPATPTRA SESPARGPSG SPRTQGRGGP ASVPSSSPGT SVKKPDPNIK NNAASEGVLA SFFNSLLSKK TGSPGSPGAG GVQSTAKNSG QKTVLSNVQE ELDRMTRKPD SMVTNSSTEN EA
Cytoplasmic dynein is a microtubule-associated motorprotein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic dynein 1 light intermediate chain 2; Dynein light intermediate chain 2, cytosolic; dynein, cytoplasmic 1, light intermediate chain 2; dynein, cytoplasmic, light intermediate polypeptide 2; LIC-2; LIC53/55
Gene Aliases: DNCLI2; DYNC1LI2; LIC2
UniProt ID: (Human) O43237
Entrez Gene ID: (Human) 1783
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support