Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LLSETLDLSE LFHPDTFLNA LRQETARAVG RSVDSLKFVA SWKGRLQEAK LQIKISGLLL EGCSFDGNQL SENQLDSPSV SSVLPCFMGW IPQDA
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic dynein 2 heavy chain; Cytoplasmic dynein 2 heavy chain 1; Dynein cytoplasmic heavy chain 2; Dynein heavy chain 11; Dynein heavy chain isotype 1B; dynein heavy chain, isotype 1B; dynein, cytoplasmic 2, heavy chain 1; dynein, cytoplasmic, heavy polypeptide 2; hDHC11
Gene Aliases: ATD3; DHC1B; DHC2; DNCH2; DYH1B; DYNC2H1; hdhc11; KIAA1997; SRPS2B; SRTD3
UniProt ID: (Human) Q8NCM8
Entrez Gene ID: (Human) 79659
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support