Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Cytoplasmic dyneins are large enzyme complexes with a molecular mass of about 1, 200 kD. They contain two force-producing heads formed primarily from dynein heavy chains, and stalks linking the heads to a basal domain, which contains a varying number of accessory intermediate chains. The complex is involved in intracellular transport and motility. DYNLL1 is a light chain and exists as part of this complex but also physically interacts with and inhibits the activity of neuronal nitric oxide synthase. Binding of this protein destabilizes the neuronal nitric oxide synthase dimer, a conformation necessary for activity, and it may regulate numerous biologic processes through its effects on nitric oxide synthase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 8 kDa dynein light chain; 8kD LC; 8kDa LC; cytoplasmic dynein light polypeptide; DLC1DNCLC1; DLC8; DLC8MGC126137; DNCL1MGC126138; dynein LC8; Dynein light chain 1, cytoplasmic; Dynein light chain LC8-type 1; dynein, cytoplasmic, light chain 1; dynein, cytoplasmic, light peptide; dynein, cytoplasmic, light polypeptide 1; dynein, light chain, LC8-type 1; H LC8; LC8-type 1; MGC126137; MGC126138; mPIN; PIN; PINLC8a; Protein inhibitor of neuronal nitric oxide synthase; protein inhibitor of nitric oxide synthase
Gene Aliases: 8kDLC; DLC1; DLC8; DNCL1; DNCLC1; DYNLL1; HDLC1; LC8; LC8a; PIN
UniProt ID: (Human) P63167, (Rat) P63170, (Mouse) P63168
Entrez Gene ID: (Human) 8655, (Rat) 58945, (Mouse) 56455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support