Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: YEVLETIGK GSFGQVAKCL DHKNNELVAL KIIRNKKRFH QQALMELKIL EALRKKDKDN TYNVVHMKDF FYFRNHFCIT FELLGINLYE LMKNNNFQGF SLSIVRRFTL SVLKCLQMLS VEKIIHCDLK PENIVLYQKG QASVKVIDFG SSCYEHQKVY TYIQSRFYRS PEVILGHPYD VAIDMWSLGC ITAELYTGYP LFPGENEVEQ LACIMEVLGL PPAGFIQTAS RRQTFFDSKG FPKNITNNRG KKRYPDSKDL TMVLKTYDTS FLDFLRRCLV WEPSLRMTPD QALKHAWI (104-400 aa encoded by BC031244)
Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 4; Dual specificity tyrosine-phosphorylation-regulated kinase 4; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4; dyrk4; RAD51 associated protein 1
Gene Aliases: AW049118; DYRK4; Dyrk4a; Dyrk4b; Rad51ap1
UniProt ID: (Human) Q9NR20, (Mouse) Q8BI55
Entrez Gene ID: (Human) 8798, (Mouse) 101320, (Rat) 312721
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support