Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPLQVSDYSW QQTKTAVFLS LPLKGVCVRD TDVFCTENYL KVNFPPFLFE AFLYAPIDDE SSKAKIGNDT IVFTLYKKEA AMWETLSVTG VDKEMMQRIR EKSILQAQER AKEATEAKAA AKREDQKYAL SVMMKIEEEE RKKIEDMKEN ERIKATKALE AWKEYQRKAE EQKKIQREEK LCQKEKQIKE GRKKIKYKSL TRNLASRNLA PKGRNSENIF TEKLKEDSIP APRSVGSIKI NFTPRVFPTA LRESQVAEEE EWLHKQAEAR RAMNTDIAEL CDLKEEEKNP EWLKDKGNKL FATENYLAAI NAYNLAIRLN NKMPLLYLNR AVCHLKLKNL HKAIEDSSKE FCSLEGIECQ ASEPKLSHHI PSDLHVYIQM A
This gene encodes a tetratricopeptide repeat domain-containing protein. The encoded protein interacts with estrogen receptors and the heat shock proteins, Hsp70 and Hsp90. A chromosomal translocation involving this gene is associated with a susceptibility to developmental dyslexia. Mutations in this gene are associated with deficits in reading and spelling. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dynein axonemal assembly factor 4; dynein, axonemal, assembly factor 4; Dyslexia susceptibility 1 candidate gene 1 protein; Dyslexia susceptibility 1 candidate gene 1 protein homolog; FLJ37882; MGC70618
Gene Aliases: 1700010I24Rik; b2b811Clo; CILD25; DNAAF4; DYX1; DYX1C1; DYXC1; EKN1; RD
UniProt ID: (Human) Q8WXU2, (Mouse) Q8R368
Entrez Gene ID: (Human) 161582, (Mouse) 67685
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support