Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • DEAF1 Antibodies

          Fabgennix

          Deformed Epidermal Autoregulatory Factor 1 Polyclonal Antibody

          View all (20) DEAF1 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Deformed Epidermal Autoregulatory Factor 1 Polyclonal Antibody

          • Antibody Testing Data (1)
          Deformed Epidermal Autoregulatory Factor 1 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Deformed Epidermal Autoregulatory Factor 1 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Deformed Epidermal Autoregulatory Factor 1 Antibody (DEAF1-101AP) in IHC

          Rat Testis- DEAF1 Primary Antibody: DEAF1-101AP; 1:50 dilution in IHC Blocking Buffer. DAB (brown) staining and Hematoxylin QS (blue) counterstain. 40X magnification. FFPE section. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Deformed Epidermal Autoregulatory Factor 1 Antibody in Immunohistochemistry (IHC)

          Product Details

          DEAF1-101AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (IHC)

          1:50-1:250
          -

          ELISA (ELISA)

          1:10,000
          -

          Immunoprecipitation (IP)

          1:50-1:250
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide taken within amino acid region 500-565 on mouse Deformed epidermal autoregulatory factor 1 homolog protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Target Information

          HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DEAF-1 related transcriptional regulator (NUDR); DEAF1 transcription factor; Deformed epidermal autoregulatory factor 1 homolog; Nuclear DEAF-1-related transcriptional regulator; NUDR; Suppressin; Zinc finger MYND domain-containing protein 5

          View more View less

          Gene Aliases: AU042387; C230009B13Rik; DEAF1; MRD24; NUDR; SPN; ZMYND5

          View more View less

          UniProt ID: (Human) O75398, (Mouse) Q9Z1T5

          View more View less

          Entrez Gene ID: (Human) 10522, (Mouse) 54006, (Rat) 83632

          View more View less

          Function(s)
          DNA binding transcription factor activity, sequence-specific DNA binding protein binding metal ion binding
          Process(es)
          behavioral fear response neural tube closure transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter multicellular organism development nervous system development visual learning regulation of mammary gland epithelial cell proliferation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter embryonic skeletal system development germ cell development anatomical structure morphogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0