Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is against the N terminal 86aa of full length p50 dynamitin.
Immunogen sequence: MADPKYADL PGIARNEPDV YETSDLPEDD QAEFDAEELT STSVEHIIVN PNAAYDKFKD KRVGTKGLDF SDRIGKTKRT GYESGEYEML GEGLGVKETP QQKYQRLLHE VQELTTEVEK IKTTVKESAT EEKLTPVLLA KQLAALKQQL VASHLEKLLG PDAAINLTDP DGALAKRLLL QLEATKNSKG GSGGKTTGTP PDSSLVTYEL HSRPEQDKFS QAAKVAELEK RLTELETAVR CDQDAQNPLS AGLQGACLME TVELLQAKVS ALDLAVLDQV EARLQSVLGK VNEIAKHKAS VEDADTQSKV HQLYETIQRW SPIASTLPEL VQRLVTIKQL HEQAMQFGQL LTHLDTTQQM IANSLKDNTT LLTQVQTTMR ENLATVEGNF ASIDERMKKL GK (1-401 aa encoded by BC014083)
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 50 kDa dynein-associated polypeptide; DCTN-50; dynactin 2 (p50); Dynactin complex 50 kDa subunit; Dynactin subunit 2; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; p50 dynamitin
Gene Aliases: 2310042E05Rik; C130077D06Rik; C87049; DCTN-50; DCTN2; DCTN50; DYNAMITIN; GMP23-48K; HEL-S-77; p50; RBP50
UniProt ID: (Human) Q13561, (Mouse) Q99KJ8
Entrez Gene ID: (Human) 10540, (Mouse) 69654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support