Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Sequence of this protein is as follows: MFGIQESIQR SGSSMKEEPL GSGMNAVRTW MQGAGVLDAN TAAQSGVGLA RAHFEKQPPS NLRKSNFFHF VLALYDRQGQ PVEIERTAFV GFVEKEKEAN SEKTNNGIHY RLQLLYSNGI RTEQDFYVRL IDSMTKQAIV YEGQDKNPEM CRVLLTHEIM CSRCCDKKSC GNRNETPSDP VIIDRFFSKF FLKCNQNCLK NAGNPRDMRR FQVVVSTTVN VDGHVLAVSD NMFVHNNSKH GRRARRLDPS EGTPSYLEHA TPCIKAISPS EGWTTGGATV IIIGDNFFDG LQVIFGTMLV WSELITPHAI RVQTPPRHIP GVVEVTLSYK SKQFCKGTPG RFIYTALNEP TIDYGFQRLQ KVIPRHPGDP ERLPKEVILK RAADLVEALY GMPHNNQEII LKRAADIAEA LYSVPRNHNQ LPALANTSVH AGMMGVNSFS GQLAVNVSEA SQATNQGFTR NSSSVSPHGY VPSTTPQQTN YNSVTTSMNG YGSAAMSNLG GSPTFLNGSA ANSPYAIVPS SPTMASSTSL PSNCSSSSGI FSFSPANMVS AVKQKSAFAP VVRPQTSPPP TCTSTNGNSL QAISGMIVPP M
Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. Target protein is a transcription factor that may regulate several olfactory-specific genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collier; Collier, Olf and EBF transcription factor 1; Early B-cell factor; O/E-1; olfactory neuronal transcription factor 1; Transcription factor COE1
Gene Aliases: COE1; EBF; EBF1; O/E-1; OLF1
UniProt ID: (Human) Q9UH73
Entrez Gene ID: (Human) 1879
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support