Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: A375, HepG2.
Immunogen sequence: SIEKISKVEC GFATIKDLRD HKLDNRMESF FLAETVKYLY LLFDPTNFIH NNGSTFDAVI TPYGECILGA GGYIFNTEAH PIDPAALHCC QRLKEEQWEV EDLMREFYSL KRSRSKFQKN TVSSGPWEPP ARPGTLFSPE NHDQARERKP AKQKVPLLSC PSQPFTSKLA LLGQVFLDSS
In the endoplasmic reticulum (ER), misfolded proteins are retrotranslocated to the cytosol and degraded by the proteasome in a process known as ER-associated degradation (ERAD). EDEM2 belongs to a family of proteins involved in ERAD of glycoproteins.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER degradation enhancer, mannosidase alpha-like 2; ER degradation-enhancing alpha-mannosidase-like 2; ER degradation-enhancing alpha-mannosidase-like protein 2; ER degradation-enhancing-mannosidase-like protein 2
Gene Aliases: 9530090G24Rik; AI327354; bA4204.1; C20orf31; C20orf49; EDEM2; UNQ573/PRO1135
UniProt ID: (Human) Q9BV94, (Mouse) Q8BJT9
Entrez Gene ID: (Human) 55741, (Mouse) 108687
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support