Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: DIRGDKITWI EGKEPGCETI GLLMSSMDDL IRHCNGKLGS YKINGRTKAM VACYPGNGTG YVRHVDNPNG DGRCVTCIYY LNKDWDAKVS GGILRIFPEG K
Cellular oxygen sensor that catalyzes, under normoxic conditions, the post-translational formation of 4-hydroxyproline in hypoxia-inducible factor (HIF) alpha proteins. Hydroxylates a specific proline found in each of the oxygen-dependent degradation (ODD) domains (N-terminal, NODD, and C-terminal, CODD) of HIF1A. Also hydroxylates HIF2A. Has a preference for the CODD site for both HIF1A and HIF1B. Hydroxylated HIFs are then targeted for proteasomal degradation via the von Hippel-Lindau ubiquitination complex. Under hypoxic conditions, the hydroxylation reaction is attenuated allowing HIFs to escape degradation resulting in their translocation to the nucleus, heterodimerization with HIF1B, and increased expression of hypoxy-inducible genes. EGLN1 is the most important isozyme under normoxia and, through regulating the stability of HIF1, involved in various hypoxia-influenced processes such as angiogenesis in retinal and cardiac functionality. Target proteins are preferentially recognized via a LXXLAP motif.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Egl nine homolog 1; Egl nine homolog 2; egl nine-like protein 1; egl-9 family hypoxia-inducible factor 1; egl-9 family hypoxia-inducible factor 2; EIT-6; Estrogen-induced tag 6; HIF prolyl hydroxylase 2; HIF-PH1; HIF-PH2; HIF-prolyl hydroxylase 1; HIF-prolyl hydroxylase 2; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; Hypoxia-inducible factor prolyl hydroxylase 2; PHD1; PHD2; Prolyl hydroxylase domain-containing protein 1; Prolyl hydroxylase domain-containing protein 2; Prolyl hydroxylase EGLN2; SM-20; zinc finger MYND domain-containing protein 6
Gene Aliases: C1orf12; ECYT3; EGLN1; EGLN2; EIT6; HALAH; HIF-PH1; HIF-PH2; HIFPH1; HIFPH2; HPH-1; HPH-2; HPH-3; HPH2; PHD1; PHD2; PNAS-118; PNAS-137; SM20; ZMYND6
UniProt ID: (Human) Q9GZT9, (Human) Q96KS0
Entrez Gene ID: (Human) 54583, (Human) 112398
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support