Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GCHYRIAHA EAQVGLPEVT LGLLPGARGT QLLPRLTGVP AALDLITSGR RILADEALKL GILDKVVNSD PVEEAIRFAQ RVSDQPLESR RLCNKPIQSL PNMDSIFSEA LLKMRRQHPG CLAQEACVRA VQAAVQYPYE VGIKKEEELF LYLLQSGQAR ALQYAFFAER KANKWSTP (108-284 aa encoded by BC038948)
As an L-hydroxy-specific enzyme, EHHADH (enoyl-CoA-hydratase:3-hydroxyacyl-CoA dehydrogenase), also known as Peroxisomal L-bifunctional enzyme, is a 723 amino acid protein has an essential tripeptide sequence on its carboxyl-terminus that is required for peroxisomal transport. EHHADH-null mice only exhibit a blunted peroxisome proliferative response when challenged with a peroxisome proliferator. Since there were no observed changes in lipid metabolism, this evidence suggests that enoyl-CoAs were diverted to the D-hydroxy-specific beta-oxidation system for metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3,2-trans-enoyl-CoA isomerase; enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-bifunctional enzyme; L-bifunctional protein; L-bifunctional protein, peroxisomal; L-PBE; L-peroxisomal bifunctional enzyme; L-specific bifunctional protein; L-specific multifunctional beta-oxdiation protein; LBP; MFE1; MFP-1; MFP1; Multifunctional enzyme 1; multifunctional enzyme type 1; Multifunctional protein 1; PBE; Peroxisomal bifunctional enzyme; peroxisomal bifunctional enzyme type 1; peroxisomal enoyl-CoA hydratase
Gene Aliases: ECHD; EHHADH; HD; LBFP; LBP; MEF; Mfe; MFP; Pbe; PBFE; 1; 1300002P22Rik; FRTS3; L-PBE; Mfe1; MFP1; perMFE-1
UniProt ID: (Human) Q08426, (Mouse) Q9DBM2, (Rat) P07896
Entrez Gene ID: (Human) 1962, (Mouse) 74147, (Rat) 171142
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support