Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: YDPIKTAQGS LSLKAYRLTP KLMEVCKEKD FSPEALKKAN ITFEYMFEEV PIVIKNSHLI NVLMWELEKK SAVADKHELL SLASSNHLGK NLQLLMDRV
eIF3H is a component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-3-gamma; eIF3 p40 subunit; EIF3H; eIF3p40; Eukaryotic translation initiation factor 3 subunit 3; Eukaryotic translation initiation factor 3 subunit H; eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD); eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa; eukaryotic translation initiation factor 3, subunit H; MGC102958
Gene Aliases: eIF3-gamma; eIF3-p40; EIF3H; EIF3S3
UniProt ID: (Human) O15372
Entrez Gene ID: (Human) 8667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support