Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MASRKEGTGS TATSSSSTAG AAGKGKGKGG SGDSAVKQVQ IDGLVVLKII KHYQEEGQGT EVVQGVLLGL VVEDRLEITN CFPFPQHTED DADFDEVQYQ MEMMRSLRHV NIDHLHVGWY QSTYYGSFVT RALLDSQFSY QHAIEESVVL IYDPIKTAQG SLSLKAYRLT PKLMEVCKEK DFSPEALKKA NITFEYMFEE VPIVIKNSHL INVLMWELEK KSAVADKHEL LSLASSNHLG KNLQLLMDRV DEMSQDIVKY NTYMRNTSKQ QQQKHQYQQR RQQENMQRQS RGEPPLPEED LSKLFKPPQP PARMDSLLIA GQINTYCQNI KEFTAQNLGK LFMAQALQEY NN
eIF3H is a component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-3-gamma; eIF3 p40 subunit; EIF3H; eIF3p40; Eukaryotic translation initiation factor 3 subunit 3; Eukaryotic translation initiation factor 3 subunit H; eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD); eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa; eukaryotic translation initiation factor 3, subunit H; MGC102958
Gene Aliases: eIF3-gamma; eIF3-p40; EIF3H; EIF3S3
UniProt ID: (Human) O15372
Entrez Gene ID: (Human) 8667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support