Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • eIF4A2 Antibodies

          Proteintech

          EIF4A2 Monoclonal Antibody (2D11E11), CoraLite® Plus 647

          View all (17) eIF4A2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite EIF4A2 Monoclonal Antibody (2D11E11), CoraLite® Plus 647

          • Antibody Testing Data (2)
          EIF4A2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          EIF4A2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EIF4A2 Antibody (CL647-67799) in Flow

          1X10^6 HEK-293 cells were intracellularly stained with 0.4 µg CoraLite® Plus 647 Anti-Human EIF4A2 (CL647-67799, Clone:2D11E11) (red), or 0.4 µg CoraLite® Plus 647 Mouse IgG2a Isotype Control (C1.18.4) (CL647-65208, Clone: C1.18.4) (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EIF4A2 Antibody in Flow Cytometry (Flow)
          EIF4A2 Antibody in Flow Cytometry (Flow)

          Product Details

          CL647-67799

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.4 µg/1x10^6 cell suspension
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2D11E11

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
          • Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation

          Excitation/Emission Max

          654/674 nm

          Form

          Liquid

          Purification

          Protein A

          Storage buffer

          PBS with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MSGGSADYNS REHGGPEGMD PDGVIESNWN EIVDNFDDMN LKESLLRGIY AYGFEKPSAI QQRAIIPCIK GYDVIAQAQS GTGKTATFAI SILQQLEIEF KETQALVLAP TRELAQQIQK VILALGDYMG ATCHACIGGT NVRNEMQKLQ AEAPHIVVGT PGRVFDMLNR RYLSPKWIKM FVLDEADEML SRGFKDQIYE IFQKLNTSIQ VVLLSATMPT DVLEVTKKFM RDPIRILVKK EELTLEGIKQ FYINVEREEW KLDTLCDLYE TLTITQAVIF LNTRRKVDWL TEKMHARDFT VSALHGDMDQ KERDVIMREF RSGSSRVLIT TDLLARGIDV QQVSLVINYD LPTNRENYIH RIGRGGRFGR KGVAINFVTE EDKRILRDIE TFYNTTVEEM PMNVADLI

          Target Information

          eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATP dependent RNA helicase eIF4A 2; ATP-dependent RNA helicase eIF4A-2; eIF-4A-II; eIF4A-II; Eukaryotic initiation factor 4A-II; eukaryotic translation initiation factor 4A isoform 2; eukaryotic translation initiation factor 4A, isoform 2; N-terminally processed; protein synthesis initiation factor 4AII

          View more View less

          Gene Aliases: 4833432N07Rik; BM-010; DDX2B; eIF-4A-II; Eif4; EIF4A; eIF4A-II; EIF4A2; EIF4F

          View more View less

          UniProt ID: (Human) Q14240, (Mouse) P10630, (Rat) Q5RKI1

          View more View less

          Entrez Gene ID: (Human) 1974, (Mouse) 13682, (Rat) 303831

          View more View less

          Function(s)
          translation initiation factor activity ATP-dependent RNA helicase activity helicase activity protein binding ATP binding ATPase activity poly(A) RNA binding nucleotide binding nucleic acid binding RNA binding hydrolase activity
          Process(es)
          nuclear-transcribed mRNA poly(A) tail shortening translational initiation regulation of translational initiation RNA secondary structure unwinding viral process negative regulation of RNA-directed RNA polymerase activity translation regulation of gene expression
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0