Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSVNVNRSV SDQFYRYKMP RLIAKVEGKG NGIKTVIVNM VDVAKALNRP PTYPTKYFGC ELGAQTQFDV KNDRYIVNGS HEANKLQDML DGFIKKFVLC PECENPETDL HVNPKKQTIG NSCKACGYRG MLDTHHKLCT FILKNPPENS DSGTGKKEKE KKNRKGKDKE NGSVSSSETP PPPPPPNEIN PPPHTMEEEE DDDWGEDTTE EAQRRRMDEI SDHAKVLTLS DDLERTIEER VNILFDFVKK KKEEGVIDSS DKEIVAEAER LDVKAMGPLV LTEVLFNEKI REQIKKYRRH FLRFCHNN (1-307 aa encoded by BC032866)
Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-5; eukaryotic initiation factor 5 (eIF-5); Eukaryotic translation initiation factor 5
Gene Aliases: 2810011H21Rik; D12Ertd549e; EIF-5; EIF-5A; EIF5
UniProt ID: (Human) P55010, (Mouse) P59325
Entrez Gene ID: (Human) 1983, (Mouse) 217869, (Rat) 56783
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support