Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TNRVPPTLN GHLNPTSEKS AAGLPLPPAA AAIPTPPPLP STYLPISHPP QIVNSNSNSP STPEGRGTQD LPVDSFSQND SIYEDQQDKY TSRTSLETLP PGSVLLKCPK PMEENHSMSH KKSKKKSKKH KEKDQIKKHD IETIEEKEED LKREEEIAKL NNSSPNSSGG VKEDCTASME PSAIELPDYL IKYIAIVSYE QRQNYKDDFN AEYDEYRALH ARMETVARRF IKLDAQRKRL SPGSKEYQNV HEEVLQEYQK IKQSSPNYHE EKYRCEYLHN KLAHIKRLIG EFDQQQAESW S (341-640 aa encoded by BC028412)
SHP-1 (PTPN6) is a non-receptor protein tyrosine phosphatase that is expressed primarily in hematopoietic cells. The enzyme is composed of two SH2 domains, a tyrosine phosphatase catalytic domain, and a carboxy-terminal regulatory domain. SHP-1 removes phosphates from target proteins to downregulate several tyrosine kinase-regulated pathways. In hematopoietic cells, the amino-terminal SH2 domain of SHP-1 binds to tyrosine phosphorylated erythropoietin receptors (EpoR) to negatively regulate hematopoietic growth. Overexpression of SHP-1 in epithelial cells results in dephosphorylation of the Ros receptor tyrosine kinase and subsequent downregulation of Ros-dependent cell proliferation and transformation. Following ligand binding in myeloid cells, SHP-1 associates with the IL-3R beta chain and downregulates IL-3-induced tyrosine phosphorylation and cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eleven-nineteen lysine-rich leukemia gene 2; ELL-related RNA polymerase II, elongation factor; elongation factor RNA for polymerase II 2; elongation factor, RNA polymerase II, 2; RNA polymerase II elongation factor ELL2
Gene Aliases: ELL2
UniProt ID: (Human) O00472, (Mouse) Q3UKU1
Entrez Gene ID: (Human) 22936, (Mouse) 192657, (Rat) 309918
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support