Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRQKRKGDLS PAELMMLTIG DVIKQLIEAH EQGKDIDLNK VKTKTAAKYG LSAQPRLVDI IAAVPPQYRK VLMPKLKAKP IRTASGIAVV AVMCKPHRCP HISFTGNICV YCPGGPDSDF EYSTQSYTGY EPTSMRAIRA RYDPFLQTRH RIEQLKQLGH SVDKVEFIVM GGTFMALPEE YRDYFIRNLH DALSGHTSNN IYEAVKYSER SLTKCIGITI ETRPDYCMKR HLSDMLTYGC TRLEIGVQSV YEDVARDTNR GHTVKAVCES FHLAKDSGFK VVAHMMPDLP NVGLERDIEQ FTEFFENPAF RPDGLKLYPT LVIRGTGLYE LWKSGRYKSY SPSDLVELVA RILALVPPWT RVYRVQRDIP MPLVSSGVEH GNLRELALAR MKDLGIQCRD VRTREVGIQE IHHKVRPYQV ELVRRDYVAN GGWETFLSYE DPDQDILIGL LRLRKCSEET FRFELGGGVS IVRELHVYGS VVPVSSRDPT KFQHQGFGML LMEEAERIAR EEHGSGKIAV ISGVGTRNYY RKIGYRLQGP YMVKMLK
Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.3.1.48; elongation protein 3 homolog; Elongator complex protein 3; h KAT9; hELP3; tRNA uridine(34) acetyltransferase
Gene Aliases: ELP3; KAT9
UniProt ID: (Human) Q9H9T3
Entrez Gene ID: (Human) 55140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support