Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Required for the assembly of a functional nuclear pore complex (NPC) on the surface of chromosomes as nuclei form at the end of mitosis. May initiate NPC assembly by binding to chromatin and recruiting the Nup107-160 subcomplex of the NPC. Also required for the localization of the Nup107-160 subcomplex of the NPC to the kinetochore during mitosis and for the completion of cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AT hook containing transcription factor 1; ELYS transcription factor like protein TMBS62; ELYS transcription factor-like protein TMBS62; Embryonic large molecule derived from yolk sac; Protein ELYS; Protein MEL-28; Putative AT-hook-containing transcription factor 1
Gene Aliases: 6230412P20Rik; AHCTF1; AV011447; ELYS; MST108; MSTP108; RGD1561291; TMBS62
UniProt ID: (Human) Q8WYP5, (Mouse) Q8CJF7
Entrez Gene ID: (Human) 25909, (Rat) 360886, (Mouse) 226747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support