Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: GSEAKNSGAS CPPCPKYASC HNSTHCTCED GFRARSGRTY FHDSSEKCED INECETGLAK CKYKAYCRNK VGG
FIRE is a 689 amino acid protein that maps to human gene EMR4. FIRE is also known as EGF-like module-containing mucin-like hormone receptor-like 4 and is a member of the the G protein-coupled receptor 2 family, LN-TM7 subfamily. The epidermal growth factor-transmembrane seven (EGF-TM7) family are proteins that express EGF-like domains at their extracellular N-terminus coupled to a classical seven-transmembrane (TM7) cassette. FIRE is believed to mediate the cellular interaction between myeloid cells and beta cells. FIRE is a membrane bound, multi-pass protein that is proteolytically cleaved into two subunits, an extracellular alpha subunit and a seven-transmembrane subunit. FIRE contains one GPS (G protein-coupled receptor proteolytic cleavage site) domain and two EGF (epidermal growth factor)-like domains, the second of which mediates the interaction with the putative ligand.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene Aliases: EMR4; EMR4P; FIRE; GPR127; PGR16
Entrez Gene ID: (Human) 326342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support