Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SILKIHAREI FDSRGNPTVE VDLFTSKGLF RAAVPSGAST GIYEALELRD NDKTRYMGKG VSKAVEHINK TIAPALVSKK LNVTEQEKID KLMIEMDGTE NKSKFGANAI LGVSLAVCKA GAVEKGVPLY RHIADLAGNS EVILPVPAFN VINGGSHAGN KLAMQEFMIL PVGAANFREA MRIGAEVYHN LKNVIKEKYG KDATNVGDEG GFAPNILENK EGLELLKTAI GKAGYTDKVV IGMDVAASEF FRSGKYDLDF KSPDDPSRYI SPDQLADLYK SFIKDYPVVS IEDPFDQDDW GAWQKFTASA GIQVVGDDLT VTNPKRIAKA VNEKSCNCLL LKVNQIGSVT ESLQACKLAQ ANGWGVMVSH RSGETEDTFI ADLVVGLCTG QIKTGAPCRS ERLAKYNQLL RIEEELGSKA KFAGRNFRNP LAK
Enolase 1 is a multifunctional enzyme that, as well as its role in glycolysis, plays a part in various processes such as growth control, hypoxia tolerance and allergic responses. May also function in the intravascular and pericellular fibrinolytic system due to its ability to serve as a receptor and activator of plasminogen on the cell surface of several cell-types such as leukocytes and neurons. Stimulates immunoglobulin production.MBP1 binds to the myc promoter and acts as a transcriptional repressor. May be a tumor suppressor.Enolase 2 has neurotrophic and neuroprotective properties on a broad spectrum of central nervous system (CNS) neurons. Binds, in a calcium-dependent manner, to cultured neocortical neurons and promotes cell survival.Enolase 3 appears to have a function in striated muscle development and regeneration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-phospho-D-glycerate hydro-lyase; 2-phospho-D-glycerate hydrolase; 2-phospho-D-glycerate hydrolyase; alpha enolase; alpha enolase like 1; Alpha-enolase; Beta-enolase; beta-enolase 3; c-Myc promoter binding protein; C-myc promoter-binding protein; Enolase 1; enolase 1, (alpha); enolase 1, alpha non-neuron; Enolase 2; enolase 2 (gamma, neuronal); Enolase 3; enolase 3 (beta, muscle); enolase-alpha; epididymis secretory protein Li 17; epididymis secretory protein Li 279; Gamma-enolase; MBP-1; MPB-1; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); Muscle-specific enolase; MYC promoter-binding protein 1; Neural enolase; neuron specific gamma enolase; Neuron-specific enolase; neuronal enriched enolase; neurone-specific enolase; NNE; Non-neural enolase; NSE; Phosphopyruvate hydratase; Plasminogen-binding protein; Skeletal muscle enolase; tau-crystallin
Gene Aliases: 0610008I15; AI837106; AL022784; BBE; D6Ertd375e; Eno-1; Eno-2; Eno-3; ENO1; ENO1L1; ENO2; ENO3; GSD13; HEL-S-17; HEL-S-279; MBP-1; MBPB1; MPB1; MSE; NNE; NSE; PPH; RNEN3
UniProt ID: (Human) P06733, (Human) P09104, (Pig) Q1KYT0, (Human) P13929, (Mouse) P17182, (Rat) P04764, (Mouse) P17183, (Rat) P07323, (Mouse) P21550, (Rat) P15429
Entrez Gene ID: (Human) 2023, (Human) 2026, (Pig) 692156, (Human) 2027, (Mouse) 13806, (Rat) 24333, (Mouse) 13807, (Rat) 24334, (Mouse) 13808, (Rat) 25438
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support