Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ENPP2 Antibodies

          Abnova

          ENPP2 Polyclonal Antibody, MaxPab™

          View all (21) ENPP2 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ENPP2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          ENPP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ENPP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ENPP2 Antibody (H00005168-B01P) in WB

          Western Blot analysis of ENPP2 expression in transfected 293T cell line by ENPP2 MaxPab polyclonal antibody. Lane 1: ENPP2 transfected lysate(94.93 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ENPP2 Antibody in Western Blot (WB)

          Product Details

          H00005168-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ENPP2 (ABM85745.1, 1 a.a. approximately 863 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MARRSSFQSC QIISLFTFAV GVNICLGFTA HRIKRAEGWE EGPPTVLSDS PWTNISGSCK GRCFELQEAG PPDCRCDNLC KSYTSCCHDF DELCLKTARG WECTKDRCGE VRNEENACHC SEDCLARGDC CTNYQVVCKG ESHWVDDDCE EIKAAECPAG FVRPPLIIFS VDGFRASYMK KGSKVMPNIE KLRSCGTHSP YMRPVYPTKT FPNLYTLATG LYPESHGIVG NSMYDPVFDA TFHLRGREKF NHRWWGGQPL WITATKQGVK AGTFFWSVVI PHERRILTIL QWLTLPDHER PSVYAFYSEQ PDFSGHKYGP FGPEMTNPLR EIDKIVGQLM DGLKQLKLHR CVNVIFVGDH GMEDVTCDRT EFLSNYLTNV DDITLVPGTL GRIRSKFSNN AKYDPKAIIA NLTCKKPDQH FKPYLKQHLP KRLHYANNRR IEDIHLLVER RWHVARKPLD VYKKPSGKCF FQGDHGFDNK VNSMQTVFVG YGPTFKYKTK VPPFENIELY NVMCDLLGLK PAPNNGTHGS LNHLLRTNTF RPTMPEEVTR PNYPGIMYLQ SDFDLGCTCD DKVEPKNKLD ELNKRLHTKG STEERHLLYG RPAVLYRTRY DILYHTDFES GYSEIFLMPL WTSYTVSKQA EVSSVPDHLT SCVRPDVRVS PSFSQNCLAY KNDKQMSYGF LFPPYLSSSP EAKYDAFLVT NMVPMYPAFK RVWNYFQRVL VKKYASERNG VNVISGPIFD YDYDGLHDTE DKIKQYVEGS SIPVPTHYYS IITSCLDFTQ PADKCDGPLS VSSFILPHRP DNEESCNSSE DESKWVEELM KMHTARVRDI EHLTSLDFFR KTSRSYPEIL TLKTYLHTYE SEI

          Target Information

          The protein encoded by this gene functions as both a phosphodiesterase, which cleaves phosphodiester bonds at the 5' end of oligonucleotides, and a phospholipase, which catalyzes production of lysophosphatidic acid (LPA) in extracellular fluids. LPA evokes growth factor-like responses including stimulation of cell proliferation and chemotaxis. This gene product stimulates the motility of tumor cells and has angiogenic properties, and its expression is upregulated in several kinds of carcinomas. The gene product is secreted and further processed to make the biologically active form. Several alternatively spliced transcript variants encoding different isoforms have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Autotaxin; autotaxin-t; AutotaxinATX; E-NPP 2; Ectonucleotide pyrophosphatase/phosphodiesterase family member 2; Extracellular lysophospholipase D; FLJ26803; LysoPLD; phosphodiesterase I/nucleotide pyrophosphatase 2; plasma lysophospholipase D

          View more View less

          Gene Aliases: ATX; ATX-X; AUTOTAXIN; ENPP2; LysoPLD; NPP2; PD-IALPHA; PDNP2

          View more View less

          UniProt ID: (Human) Q13822

          View more View less

          Entrez Gene ID: (Human) 5168

          View more View less

          Function(s)
          nucleic acid binding phosphodiesterase I activity nucleotide diphosphatase activity lysophospholipase activity scavenger receptor activity calcium ion binding transcription factor binding zinc ion binding hydrolase activity polysaccharide binding alkylglycerophosphoethanolamine phosphodiesterase activity
          Process(es)
          phosphate-containing compound metabolic process receptor-mediated endocytosis movement of cell or subcellular component chemotaxis immune response G-protein coupled receptor signaling pathway phospholipid catabolic process positive regulation of epithelial cell migration regulation of cell migration phosphatidylcholine catabolic process regulation of angiogenesis positive regulation of peptidyl-tyrosine phosphorylation nucleic acid phosphodiester bond hydrolysis positive regulation of lamellipodium morphogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0