Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E-NPP 5; E-NPP5; EC 3.1; Ectonucleotide pyrophosphatase/phosphodiesterase family member 5; ENPP5; KIAA0879; NPP-5
Gene Aliases: AW061004; D17Abb1e; E-NPP 5; ENPP5; mKIAA4152; NPP-5; NPP5; UNQ550/PRO1107
UniProt ID: (Human) Q9UJA9, (Mouse) Q9EQG7, (Rat) P84039
Entrez Gene ID: (Human) 59084, (Mouse) 83965, (Rat) 316249
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support