Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, no cross-reactivity with recombinant human (rh) ENPP-2, rhENPP-7, or recombinant mouse ENPP-7 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E-NPP 5; EC 3.1; Ectonucleotide pyrophosphatase/phosphodiesterase family member 5; ENPP5; KIAA0879
Gene Aliases: ENPP5; NPP-5; NPP5; UNQ550/PRO1107
UniProt ID: (Human) Q9UJA9
Entrez Gene ID: (Human) 59084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support