Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HIF-2 alpha Antibodies

          Abnova

          EPAS1 Polyclonal Antibody, MaxPab™

          View all (36) HIF-2 alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite EPAS1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          EPAS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          EPAS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EPAS1 Antibody (H00002034-B02P) in WB

          Western Blot analysis of EPAS1 expression in transfected 293T cell line by EPAS1 MaxPab polyclonal antibody. Lane 1: EPAS1 transfected lysate(96.50 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EPAS1 Antibody in Western Blot (WB)

          Product Details

          H00002034-B02P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EPAS1 (NP_001421.2, 1 a.a. approximately 870 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MTADKEKKRS SSERRKEKSR DAARCRRSKE TEVFYELAHE LPLPHSVSSH LDKASIMRLA ISFLRTHKLL SSVCSENESE AEADQQMDNL YLKALEGFIA VVTQDGDMIF LSENISKFMG LTQVELTGHS IFDFTHPCDH EEIRENLSLK NGSGFGKKSK DMSTERDFFM RMKCTVTNRG RTVNLKSATW KVLHCTGQVK VYNNCPPHNS LCGYKEPLLS CLIIMCEPIQ HPSHMDIPLD SKTFLSRHSM DMKFTYCDDR ITELIGYHPE ELLGRSAYEF YHALDSENMT KSHQNLCTKG QVVSGQYRML AKHGGYVWLE TQGTVIYNPR NLQPQCIMCV NYVLSEIEKN DVVFSMDQTE SLFKPHLMAM NSIFDSSGKG AVSEKSNFLF TKLKEEPEEL AQLAPTPGDA IISLDFGNQN FEESSAYGKA ILPPSQPWAT ELRSHSTQSE AGSLPAFTVP QAAAPGSTTP SATSSSSSCS TPNSPEDYYT SLDNDLKIEV IEKLFAMDTE AKDQCSTQTD FNELDLETLA PYIPMDGEDF QLSPICPEER LLAENPQSTP QHCFSAMTNI FQPLAPVAPH SPFLLDKFQQ QLESKKTEPE HRPMSSIFFD AGSKASLPPC CGQASTPLSS MGGRSNTQWP PDPPLHFGPT KWAVGDQRTE FLGAAPLGPP VSPPHVSTFK TRSAKGFGAR GPDVLSPAMV ALSNKLKLKR QLEYEEQAFQ DLSGGDPPGG STSHLMWKRM KNLRGGSCPL MPDKPLSANV PNDKFTQNPM RGLGHPLRHL PLPQPPSAIS PGENSKSRFP PQCYATQYQD YSLSSAHKVS GMASRLLGPS FESYLLPELT RYDCEVNVPV LGSSTLLQGG DLLRALDQAT

          Target Information

          HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; Endothelial PAS domain-containing protein 1; EPAS-1; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF-2-alpha; HIF2-alpha; HLF; hypoxia-inducible factor 2 alpha; Hypoxia-inducible factor 2-alpha; Member of PAS protein 2; PAS domain-containing protein 2

          View more View less

          Gene Aliases: BHLHE73; ECYT4; EPAS1; HIF2A; HLF; MOP2; PASD2

          View more View less

          UniProt ID: (Human) Q99814

          View more View less

          Entrez Gene ID: (Human) 2034

          View more View less

          Function(s)
          RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding DNA binding protein binding transcription factor binding histone acetyltransferase binding sequence-specific DNA binding protein heterodimerization activity
          Process(es)
          angiogenesis response to hypoxia embryonic placenta development blood vessel remodeling regulation of heart rate transcription from RNA polymerase II promoter mitochondrion organization signal transduction visual perception erythrocyte differentiation lung development somatic stem cell population maintenance norepinephrine metabolic process surfactant homeostasis regulation of transcription from RNA polymerase II promoter in response to oxidative stress positive regulation of transcription from RNA polymerase II promoter cell maturation myoblast fate commitment iron ion homeostasis regulation of transcription from RNA polymerase II promoter in response to hypoxia cellular response to hypoxia
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0