Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MERLQKQPLT SPGSVSPSRD SSVPGSPSSI VAKMDNQVLG YKDLAAIPKD KAILDIERPD LMIYEPHFTY SLLEHVELPR SRERSLSPKS TSPPPSPEVW ADSRSPGIIS QASAPRTTGT PRTSLPHFHH PETSRPDSNI YKKPPIYKQR ESVGGSPQTK HLIEDLIIES SKFPAAQPPD PNQPAKIETD YWPCPPSLAV VETEWRKRKA SRRGAEEEEE EEDDDSGEEM KALRERQREE LSKVTSNLGK MILKEEMEKS LPIRRKTRSL PDRTPFHTSL HQGTSKSSSL PAYGRTTLSR LQSTEFSPSG SETGSPGLQI YPYEMLVVTN KGRTKLPPGV DRMRLERHLS AEDFSRVFAM SPEEFGKLAL WKRNELKKKA SLF
The protein encoded by this gene is an actin binding and bundling protein that plays a structural role in erythrocytes, by stabilizing and attaching the spectrin/actin cytoskeleton to the erythrocyte membrane in a phosphorylation-dependent manner. This protein contains a core domain in the N-terminus, and a headpiece domain in the C-terminus that binds F-actin. When purified from erythrocytes, this protein exists as a trimer composed of two 48 kDa polypeptides and a 52 kDa polypeptide. The different subunits arise from alternative splicing in the 3' coding region, where the headpiece domain is located. Disruption of this gene has been correlated with the autosomal dominant Marie Unna hereditary hypotrichosis disease, while loss of heterozygosity of this gene is thought to play a role in prostate cancer progression. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dematin; Dematin actin-binding protein; Erythrocyte membrane protein band 4.9; erythrocyte membrane protein band 4.9 (dematin); erythrocyte protein band 4.9
Gene Aliases: DMT; DMTN; Epb4.9; EPB49
UniProt ID: (Human) Q08495
Entrez Gene ID: (Human) 2039, (Rat) 361069
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support