Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1110021C24Rik; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; LIM domain and actin-binding protein 1; mEPLIN; sterol regulatory element binding protein 3
Gene Aliases: D15Ertd366e; EPLIN; LIMA1; PP624; SREBP3
UniProt ID: (Human) Q9UHB6, (Mouse) Q9ERG0
Entrez Gene ID: (Human) 51474, (Mouse) 65970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support