Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: DLVVVLVDVS DKWTRNQLSP QLLRCLTKYS QIPSVLVMNK VDCLKQKSVL LELTAALTEG VVNGKKLKMR QAFHSHPGTH CPSPAVKDPN TQSVGNPQRI GWPHFKEIFM LSALSQEDVK TLKQYLLTQA QPGPWEYHSA VLTSQTPEEI CANIIREKLL EHLPQEVPYN VQQKTAVWEE GPGGELVIQQ KLLVPKESYV KLLIGPKGHV ISQIAQEAGH DLMDIFLCDV DIRLSVKLLK; Positive Samples: SKOV3, HeLa, U-87MG, NCI-H460, Mouse kidney, Mouse heart, Rat liver; Cellular Location: Mitochondrion inner membrane, Mitochondrion matrix, Peripheral membrane protein
The protein encoded by this gene is a GTPase that localizes to the mitochondrion. The encoded protein binds to the 3' terminal stem loop of 12S mitochondrial rRNA and is required for proper assembly of the 28S small mitochondrial ribosomal subunit. Deletion of this gene has been shown to cause mitochondrial dysfunction, growth retardation, and apoptosis. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CEGA; Conserved ERA-like GTPase; era (E. coli G-protein homolog)-like 1; Era G-protein-like 1; Era-like 12S mitochondrial rRNA chaperone 1; ERA-like protein 1; ERA-W; GTP-binding protein era homolog; GTPase Era, mitochondrial; GTPase, human homolog of E. coli essential cell cycle protein Era; H-ERA
Gene Aliases: CEGA; ERA; ERA-W; ERAL1; ERAL1A; H-ERA; HERA; HERA-A; HERA-B
UniProt ID: (Human) O75616
Entrez Gene ID: (Human) 26284
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support