Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GHLAVYYRG GALLTSPSGP GYHIMLPFIT TFRSVQTTLQ TDEVKNVPCG TSGGVMIYID RIEVVNMLAP YAVFDIVRNY TADYDKTLIF NKIHHELNQF CSAHTLQEVY IELFDQIDEN LKQALQKDLN LMAPGLTIQA VRVTKPKIPE AIRRNFELME AEKTKLLIAA QKQKVVEKEA ETERKKAVIE AEKIAQVAKI RFQQKVMEKE TEKRISEIED AAFLAREKAK ADAEYYAAHK YATSNKHKLT PEYLELKKYQ AIASNSKIYF GSNIPNMFVD SSCALKYSDI RTGRESSLPS KEALEPSGEN VIQNKESTG (29-346 aa encoded by BC031791)
Component of the ERLIN1/ERLIN2 complex which mediates the endoplasmic reticulum-associated degradation (ERAD) of inositol 1,4,5-trisphosphate receptors (IP3Rs). Involved in regulation of cellular cholesterol homeostasis by regulation the SREBP signaling pathway. Binds cholesterol and may promote ER retention of the SCAP-SREBF complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Band_7 23-211 Keo4 (Interim) similar to C.elegans protein C42C1.9; Endoplasmic reticulum lipid raft-associated protein 1; Erlin-1; OTTHUMP00000059425; OTTHUMP00000197867; Protein KE04; Protein KE04 homolog; SPFH domain family, member 1; SPFH domain-containing protein 1; Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1
Gene Aliases: 2810439N09Rik; C10orf69; C80197; Erlin-1; ERLIN1; KE04; KEO4; RGD1307058; SPFH1; SPG62
UniProt ID: (Human) O75477, (Mouse) Q91X78
Entrez Gene ID: (Human) 10613, (Rat) 293939, (Mouse) 226144
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support